missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Candle with Glass Tube, Conical Shape

Filter Candle with narrow glass tube.

Brand:  ROBU VitraPOR™ 18311

Additional Details : Weight : 0.15680kg

 View more versions of this product

Product Code. 16862966

  • 1400.00 SEK / Pack of 2
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
Glass Tube, Ø 20 mm
65 mm
1 μm
1
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers