Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ 488 into mouse IgG1 primary antibodies.
Brand: Proteintech™ KFA021-10
This item is not returnable.
View return policy
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your mouse IgG1 antibody in <10 min without any additional equipment
Specifications
10 μL CoraLite™ 488 FlexLinker for Mouse IgG1, 20 μL FlexQuencher for Mouse IgG1, 100 μL FlexBuffer, Manual, Protocol Magnet | |
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
IgG1 |
10 x 0.5 μL Reactions | |
500 nm/520 nm | |
Mouse |