missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1
This Antibody Labeling Kit conjugates CoraLite™ 488 into mouse IgG1 primary antibodies.
1021.37 SEK - 10088.86 SEK
Specifications
Content And Storage | 1 Year at -20°C or 6 Months at +4°C, Avoid Light |
---|---|
Excitation/Emission | 500 nm/520 nm |
Labeling Target | IgG1 |
Species | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17761519
|
Proteintech™
KFA021-10 |
10 x 0.5 μL Reactions |
1021.37 SEK
10 reactions |
In Stock
Log in to see stock. |
|||||
17781519
|
Proteintech™
KFA021-50 |
50 x 0.5 μL Reactions |
4755.04 SEK
50 reactions |
In Stock
Log in to see stock. |
|||||
17771519
|
Proteintech™
KFA021-200 |
200 x 0.5 μL Reactions |
10088.86 SEK
200 reactions |
In Stock
Log in to see stock. |
|||||
Description
FlexAble CoraLite™ 488 Antibody Labeling Kit for Mouse IgG1 is a novel antibody labeling kit that uses an affinity linker to conjugate CoraLite™ 488 in any buffer condition to mouse IgG1 primary antibodies from any supplier.
- One labeling reaction requires only 0.5 μg of antibody, regardless of the antibody concentration
- Label your mouse IgG1 antibody in <10 min without any additional equipment
Specifications
1 Year at -20°C or 6 Months at +4°C, Avoid Light | |
500 nm/520 nm | |
IgG1 | |
Mouse |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title