Learn More
Flotillin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA530033
Product Code. 13454477
  • 4910.00 SEK / 100µL
Explore more special offers
Description
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (91%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. This gene encodes a caveolae-associated, integral membrane protein, which is thought to function in neuronal signaling.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Title
Flotillin 2 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated