Learn More
Euro Protection™ Freedite High-Cut Safety Shoes
Euro Protection™ Freedite High-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.
Brand:  Euro Protection™ 9FREH37
 View more versions of this product
Product Code. 15775615
  • 533.00 SEK / 1 pair
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Description
  • Footbridge for torsional rigidity.
  • Padding for snug fit.
  • Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
  • Full grain leather upper, reflective trim at the back.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
  • Composite toe cap.
  • Non-metallic (composite fabric) puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.
Specifications
Specifications
Black
Leather
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
High
602 g per shoe in size 8
Breathable 3D mesh lining
37
Polyurethane, Composite Fabric
Composite
EN ISO 20345 : 2011