Learn More
Euro Protection™ Freedite High-Cut Safety Shoes
Euro Protection™ Freedite High-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.
502.00 SEK - 533.00 SEK
Specifications
Color | Black |
---|---|
Liner Material | Breathable 3D mesh lining |
Material | Leather |
Sole Features | Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. |
Sole Material | Polyurethane, Composite Fabric |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15765615
|
Euro Protection™
9FREH36 |
36 |
533.00 SEK
1 pair |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15775615
|
Euro Protection™
9FREH37 |
37 |
533.00 SEK
1 pair |
Estimated Shipment: 28-11-2024
Log in to see stock. |
|||||
15785615
|
Euro Protection™
9FREH38 |
38 |
533.00 SEK
1 pair |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15795615
|
Euro Protection™
9FREH39 |
39 |
533.00 SEK
1 pair |
Estimated Shipment: 22-11-2024
Log in to see stock. |
|||||
15705625
|
Euro Protection™
9FREH40 |
40 |
502.00 SEK
1 pair |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15715625
|
Euro Protection™
9FREH41 |
41 |
533.00 SEK
1 pair |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15725625
|
Euro Protection™
9FREH42 |
42 |
533.00 SEK
1 pair |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
15735625
|
Euro Protection™
9FREH43 |
43 |
533.00 SEK
1 pair |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15745625
|
Euro Protection™
9FREH44 |
44 |
533.00 SEK
1 pair |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15755625
|
Euro Protection™
9FREH45 |
45 |
533.00 SEK
1 pair |
Estimated Shipment: 12-12-2024
Log in to see stock. |
|||||
15765625
|
Euro Protection™
9FREH46 |
46 |
533.00 SEK
1 pair |
Estimated Shipment: 25-11-2024
Log in to see stock. |
|||||
15775625
|
Euro Protection™
9FREH47 |
47 |
533.00 SEK
1 pair |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
15785625
|
Euro Protection™
9FREH48 |
48 |
533.00 SEK
1 pair |
Estimated Shipment: 29-11-2024
Log in to see stock. |
|||||
Beschreibung
- Footbridge for torsional rigidity.
- Padding for snug fit.
- Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
- Full grain leather upper, reflective trim at the back.
- Breathable 3D mesh lining.
- Removable, textile and foam insole.
- Composite toe cap.
- Non-metallic (composite fabric) puncture resistant sole.
- Injected polyurethane midsole.
- Polyurethane outsole.
Spezifikation
Black | |
Leather | |
Polyurethane, Composite Fabric | |
Composite | |
EN ISO 20345 : 2011 |
Breathable 3D mesh lining | |
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole. | |
High | |
602 g per shoe in size 8 |