Learn More
Euro Protection™ Freedite High-Cut Safety Shoes
Euro Protection™ Freedite High-Cut Safety Shoes are padded for a comfortable, snug fit and feature a footbridge for rigidity and a composite toe cap for safety.
502.00 SEK - 533.00 SEK
Specifications
Color Black
Liner Material Breathable 3D mesh lining
Material Leather
Sole Features Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
Sole Material Polyurethane, Composite Fabric
View More Specs
Products 13
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15765615
View Documents Promotion Details
Euro Protection™
9FREH36
36
533.00 SEK
1 pair
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
15775615
View Documents Promotion Details
Euro Protection™
9FREH37
37
533.00 SEK
1 pair
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
15785615
View Documents Promotion Details
Euro Protection™
9FREH38
38
533.00 SEK
1 pair
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
15795615
View Documents Promotion Details
Euro Protection™
9FREH39
39
533.00 SEK
1 pair
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
15705625
View Documents Promotion Details
Euro Protection™
9FREH40
40
502.00 SEK
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Add to basket
15715625
View Documents Promotion Details
Euro Protection™
9FREH41
41
533.00 SEK
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Zum Warenkorb hinzufügen
15725625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH42
42
533.00 SEK
1 pair
Estimated Shipment: 29-11-2024
to see stock.
Zum Warenkorb hinzufügen
15735625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH43
43
533.00 SEK
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Zum Warenkorb hinzufügen
15745625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH44
44
533.00 SEK
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Zum Warenkorb hinzufügen
15755625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH45
45
533.00 SEK
1 pair
Estimated Shipment: 12-12-2024
to see stock.
Zum Warenkorb hinzufügen
15765625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH46
46
533.00 SEK
1 pair
Estimated Shipment: 25-11-2024
to see stock.
Zum Warenkorb hinzufügen
15775625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH47
47
533.00 SEK
1 pair
Estimated Shipment: 05-12-2024
to see stock.
Zum Warenkorb hinzufügen
15785625
missing translation for 'viewDocuments' Promotion Details
Euro Protection™
9FREH48
48
533.00 SEK
1 pair
Estimated Shipment: 29-11-2024
to see stock.
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Footbridge for torsional rigidity.
  • Padding for snug fit.
  • Also available with an electrical hazards resistant outsole on request (Reference: 9FEHH).
  • Full grain leather upper, reflective trim at the back.
  • Breathable 3D mesh lining.
  • Removable, textile and foam insole.
  • Composite toe cap.
  • Non-metallic (composite fabric) puncture resistant sole.
  • Injected polyurethane midsole.
  • Polyurethane outsole.
Spezifikation
Spezifikation
Black
Leather
Polyurethane, Composite Fabric
Composite
EN ISO 20345 : 2011
Breathable 3D mesh lining
Removable, textile and foam insole, non-metallic (composite fabric) puncture-resistant sole, injected polyurethane midsole, polyurethane outsole.
High
602 g per shoe in size 8
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate