Learn More
Fritsch™ Stainless Steel Grinding Bowls
POA
Specifications
Product Type Grinding Bowl
Includes Lid and seal ring
Material Stainless Steel
Sample Type Medium-hard, brittle
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
13147574
Encompass
View Documents
Fritsch™
50.2100.00
250 mL
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
13121814
Encompass
View Documents
Fritsch™
50.1100.00
500 mL
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Grinding Bowl
Lid and seal ring
Stainless Steel
Medium-hard, brittle