missing translation for 'onlineSavingsMsg'
Learn More

Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

684.00 SEK - 5900.00 SEK

Chemical Identifiers

CAS 112372-14-2
Molecular Formula C8H5NO3
Molecular Weight (g/mol) 163.13
MDL Number MFCD11588128
InChI Key VHRCHODSRLMXFM-UHFFFAOYSA-N
Synonym furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
PubChem CID 13803071
IUPAC Name furo[3,2-b]pyridine-2-carboxylic acid
SMILES OC(=O)C1=CC2=NC=CC=C2O1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15407347
SDS View Documents
Thermo Scientific Alfa Aesar
H34400.MD
250 mg N/A
N/A
15487337
SDS View Documents
Thermo Scientific Alfa Aesar
H34400.03
1 g N/A
N/A
15497337
SDS View Documents
Thermo Scientific Alfa Aesar
H34400.06
5 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112372-14-2
163.13
VHRCHODSRLMXFM-UHFFFAOYSA-N
13803071
OC(=O)C1=CC2=NC=CC=C2O1
C8H5NO3
MFCD11588128
furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
furo[3,2-b]pyridine-2-carboxylic acid

Specifications

112372-14-2
C8H5NO3
furo 3,2-b pyridine-2-carboxylic acid, acmc-20mg3q, 2-carboxyfuro 3,2-b pyridine
OC(=O)C1=CC2=NC=CC=C2O1
163.13
163.13
Furo[3,2-b]pyridine-2-carboxylic acid
>300°C
MFCD11588128
VHRCHODSRLMXFM-UHFFFAOYSA-N
furo[3,2-b]pyridine-2-carboxylic acid
13803071
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Furo[3,2-b]pyridine-2-carboxylic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Furo[3,2-b]pyridine-2-carboxylic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Furo[3,2-b]pyridine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.