missing translation for 'onlineSavingsMsg'
Learn More
Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™ Product Code.: 15467415

Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™

Product Code. 15467415
5g
Click to view available options
Quantity:
1g
5g
25g
This item is not returnable. View return policy

Product Code. 15467415

Brand: Thermo Scientific Alfa Aesar 047148.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13450-91-4
Molecular Formula GaI3
Molecular Weight (g/mol) 450.436
MDL Number MFCD00016107
InChI Key DWRNSCDYNYYYHT-UHFFFAOYSA-K
Synonym gallium iii iodide, gallium iodide, gallium triiodide, acmc-20akty, gallium iodide,, gallium iii iodide, anhydrous, gallium iii iodide, ultra dry, gallium iii iodide trace metals basis, gallium iii iodide, anhydrous, powder trace metals basisShow MoreShow Less
PubChem CID 83478
IUPAC Name triiodogallane
SMILES [Ga](I)(I)I

Specifications

Quantity 5g
UN Number UN3260
Solubility Information Decomposes in water.
Formula Weight 450.43
Percent Purity 99.999%
Packaging Ampule
Sensitivity Light and moisture sensitive
Density 4.15 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) iodide, Ultra dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN02K052 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number047148.03,047148.14,047148.06
Lot NumberP11I055 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number047148.03,047148.14,047148.06
Lot NumberI25Z008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number047148.03,047148.14,047148.06
Lot NumberU10H060 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number047148.03,047148.14,047148.06
Lot NumberR25D062 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number047148.03,047148.14,047148.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gallium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.