Learn More
Lenz Laborglasintrumente Gas Washing Bottle Heads, with Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05510410
Additional Details : Weight : 0.01000kg
Description
- Acc. to DIN 12596
- With cone NS 29/32
- For bottles acc. to DIN 12463
- Made of DURAN™ tubing
Specifications
Duran Glass | |
0 |