Learn More
Lenz Laborglasintrumente Gas Washing Bottle Heads, with Filter Disc
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
504.56 SEK - 644.48 SEK
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Porosity | Price | Quantity & Availability | |||||
10603052
|
Lenz Laborglasintrumente
05530301 |
504.56 SEK
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
||||||
11516934
|
Lenz Laborglasintrumente
05510412 |
2 |
644.48 SEK
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
11596924
|
Lenz Laborglasintrumente
05510410 |
0 |
644.48 SEK
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12596
- With cone NS 29/32
- For bottles acc. to DIN 12463
- Made of DURAN™ tubing