missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eurofins DiscoverX GCN2 Protein, Active
Active, N-terminal GST-tagged, recombinant, human GCN2 full length, for use in Kinase Assays.
5796.39 SEK - 44240.30 SEK
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 215 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal GST |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17340814
|
Eurofins DiscoverX
14-934 |
10 μg |
5796.39 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17350814
|
Eurofins DiscoverX
14-934M |
250 μg |
44240.30 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GCN2 (EIF2AK4) belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1) to downregulate protein synthesis in response to varied cellular stresses.
GenPept NP_001013725
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal GST | |
Full-length | |
GCN2, active |
215 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥66% |