Learn More
Fisherbrand™ Gel Casting Gate
With integral silicone seals to effectively seal tray.
Brand: Fisherbrand™ HU20-CG
This item is not returnable.
View return policy
Description
- Casting gates
- Integral silicone seals effectively seal the tray without the need for tape
Specifications
Maxi Standard | |
Gel Casting Gate | |
Integral Silicone Seals |