Learn More
Trajan Scientific and Medical™ Glass Lined Tubing
Glass Lined Tubing (GLT) is borosilicate glass lined stainless steel tubing giving excellent resistance to the harshest of conditions
Brand:  Trajan Scientific and Medical™ 082707
 View more versions of this product
Product Code. 13120753
  • 510.00 SEK / 300mm
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Excellent reistance to strong acids and bases
  • Ideal for HPLC columns - particularly for biological matrices
  • Temperature stable up to 500°C
Specifications
Specifications
Tubing
0.3 mm
30 cm
500°C
1/16 in.
1.59 mm
Stainless Steel, Borosilicate Glass (Lining)