Learn More
Trajan Scientific and Medical™ Glass Lined Tubing
Glass Lined Tubing (GLT) is borosilicate glass lined stainless steel tubing giving excellent resistance to the harshest of conditions
Brand: Trajan Scientific and Medical™ 082717
This item is not returnable.
View return policy
Description
- Excellent reistance to strong acids and bases
- Ideal for HPLC columns - particularly for biological matrices
- Temperature stable up to 500°C
Specifications
Tubing | |
0.5 mm | |
30 cm | |
500°C |
1/16 in. | |
1.59 mm | |
Stainless Steel, Borosilicate Glass (Lining) |