Learn More
Guanosine, 98%, Thermo Scientific Chemicals
Description
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Guanosine is used as a constituent of nucleic acids. It is used in metallic paints, simulated pearls,plastics,cosmetics industry etc,. It has been also used in pharmacokinetics as a prodrug. Guanosine is used in cell culture applications as a precursor of GMP.
Solubility
Soluble in Formic Acid:Water (1:1): 50 mg/mL. Insoluble in methanol, diethyl ether.
Notes
Store away from extreme heat. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | ∼250°C (decomposition) |
Quantity | 100 g |
Beilstein | 625911 |
Merck Index | 14,4566 |
Solubility Information | Soluble in Formic Acid:Water (1:1): 50mg/mL. Insoluble in methanol,diethyl ether. |
Optical Rotation | −72° (c=1.5 in 0.1N NaOH) |
IUPAC Name | 2-amino-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one |
Formula Weight | 283.24 |
Percent Purity | ≥98% |
Chemical Name or Material | Guanosine |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10243901 | Certificate of Analysis | 08/11/2022 | A11328.18,A11328.14,A11328.22,A11328.30 |
10238001 | Certificate of Analysis | 12/04/2022 | A11328.18,A11328.14,A11328.22,A11328.30 |
10215187 | Certificate of Analysis | 22/11/2021 | A11328.18,A11328.14,A11328.22,A11328.30 |
10099306 | Certificate of Analysis | 22/11/2021 | A11328.18,A11328.14,A11328.22,A11328.30 |
10165186 | Certificate of Analysis | 22/11/2021 | A11328.18,A11328.14,A11328.22,A11328.30 |
Safety and Handling

- Guanosine
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- MIXTURE LIST-Contains : Guanosine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.