missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hematein, Thermo Scientific Chemicals
989.98 SEK - 3261.91 SEK
Chemical Identifiers
CAS | 475-25-2 |
---|---|
Molecular Formula | C16H12O6 |
Molecular Weight (g/mol) | 300.27 |
MDL Number | MFCD00036187 |
InChI Key | HLUCICHZHWJHLL-UHFFFAOYNA-N |
Synonym | hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein |
PubChem CID | 10138 |
IUPAC Name | 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one |
SMILES | OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10170952
|
Thermo Scientific Acros
370840050 |
5 g | Glass bottle |
989.98 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10222862
|
Thermo Scientific Acros
370840250 |
25 g | Glass bottle |
3261.91 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
475-25-2 | |
300.27 | |
HLUCICHZHWJHLL-UHFFFAOYNA-N | |
10138 | |
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O |
C16H12O6 | |
MFCD00036187 | |
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein | |
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one |
Specifications
475-25-2 | |
Brown | |
C16H12O6 | |
18,IV,3343 | |
hematein, haematein, hematine, hydroxybrasilein, benz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxy, 3,4,6a,10-tetrahydroxy-6,7-dihydroindeno 2,1-c chromen-9-one, 3,4,6a,10-tetrahydroxy-6a,7-dihydrobenz b indeno 1,2-d pyran-9 6h-one, 6a,7-dihydro-3,4,6a,10-tetrahydroxybenz b indeno 1,2-d pyran-9 6h-one, hematein,haematein,hydroxybrazilein,hydroxybrasilein,c.i.75290, hydroxybrazilein | |
HLUCICHZHWJHLL-UHFFFAOYNA-N | |
3,4,6a,10-tetrahydroxy-6,7-dihydroindeno[2,1-c]chromen-9-one | |
10138 | |
Crystalline Powder |
180°C | |
Authentic | |
MFCD00036187 | |
15,4669 | |
Solubility in water: very slightly soluble. Other solubilities: slightly solubel in ethanol,ether,soluble in alkaline solutions,insoluble in benzene,chloroform | |
OC1=CC=C2C3=C4C=C(O)C(=O)C=C4CC3(O)COC2=C1O | |
300.27 | |
300.26 | |
Hematein |
Safety and Handling
missing translation for 'einecsNumber' : 207-492-8
Product Identifier
- Hematein
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hematein, Thermo Scientific Chemicals