missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Hemispheric Dishes with Spout, Translucent

Round, capsule, bowl-shaped dishes with spouts constructed of translucent, durable, resistant, and chemically inert quartz glass.

Brand:  Helios Italquartz 61F00101

 View more versions of this product

Product Code. 15848004

  • 880.00 SEK / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Translucent
  • Round bottom
  • Includes spout
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications

Specifications

Translucent, includes spout
5 cc
Round
Spout
38 mm
Round
Quartz
Hemispheric Dish
14 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers