missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Hemispheric Dishes with Spout, Translucent

Round, capsule, bowl-shaped dishes with spouts constructed of translucent, durable, resistant, and chemically inert quartz glass.

Brand:  Helios Italquartz 61F00108

 View more versions of this product

Product Code. 15858004

  • 3070.00 SEK / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Translucent
  • Round bottom
  • Includes spout
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications

Specifications

Translucent, includes spout
240 cc
Round
Spout
112 mm
Round
Quartz
Hemispheric Dish
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers