Learn More
HLA-DQA2 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812936
Encompass_Preferred
 View more versions of this product
Product Code. 17567534
  • 6234.27 SEK / 100µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
HLA-DQA2 Monoclonal antibody specifically detects HLA-DQA2 in Human samples. It is validated for Western Blot
Specifications
Specifications
HLA-DQA2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P01906
Mouse
Protein A/G
RUO
3118
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI4C9
Unconjugated
HLA-DQA2
DQ alpha 2; DX ALPHA; DX alpha chain; DX-ALPHA; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA DXA; HLA-DQA1; HLA-DQA2; HLA-DXA; major histocompatibility complex, class II, DQ alpha 2; MHC class II DQA2
Human recombinant protein fragment corresponding to amino acids 24-125 of human HLA-DQA2 (NP_064440) produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
HLA-DQA2 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene