missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DQA2 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody

2789.38 SEK - 6234.27 SEK
Specifications
Antigen | HLA-DQA2 |
---|---|
Clone | OTI4C9 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17587534
![]() |
Origene Technologies
TA812936S |
30 μL |
2789.38 SEK
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17567534
![]() |
Origene Technologies
TA812936 |
100 μL |
6234.27 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-DQA2 Monoclonal antibody specifically detects HLA-DQA2 in Human samples. It is validated for Western BlotSpecifications
HLA-DQA2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
DQ alpha 2; DX ALPHA; DX alpha chain; DX-ALPHA; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA DXA; HLA-DQA1; HLA-DQA2; HLA-DXA; major histocompatibility complex, class II, DQ alpha 2; MHC class II DQA2 | |
Human recombinant protein fragment corresponding to amino acids 24-125 of human HLA-DQA2 (NP_064440) produced in E.coli. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
HLA-DQA2 |
OTI4C9 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P01906 | |
3118 | |
IgG1 | |
Protein A/G | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
HLA-DQA2 Monoclonal Antibody (OTI4C9), TrueMAB™, OriGene