missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark Professional™ Hostess™ Toilet Tissue Rolls
Dissolves in cold water so it won’t block drains. Hostess™ Small Toilet Tissue Rolls are manufactured from 100% recycled materials and are fully biodegradable.
Brand: Kimberly-Clark Professional™ 8653
402.35 SEK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- White perforated toilet tissue
- Supplied in packs of 36.
- 100% recycled
- Fully biodegradable to reduce the risk of blocked toilets
- ISO 9001 and ISO 14001 and EU Ecolabel certified
Specifications
Toilet Tissue | |
11.5 cm | |
9.4 cm | |
ISO 9001 and ISO 14001, EU Ecolabel certified | |
11.5 x 9.4 cm (4.5 x 3.7 in.) | |
18 x 2 Rolls x 320 Sheets |
4.5 in. | |
3.7 in. | |
Paper | |
White | |
Ideal for small, office and hotel washrooms, especially where economy is important | |
2-ply |