Learn More
HSD17B1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25334-1-AP-150UL
This item is not returnable.
View return policy
Description
The HSD17B1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human HSD17B1. This antibody recognizes human antigen. The HSD17B1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
HSD17B1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
3292 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human |