missing translation for 'onlineSavingsMsg'
Learn More

HSD17B1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen HSD17B1
Concentration 200 μg/mL
Applications Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883915
View Documents
Proteintech
25334-1-AP-20UL
20 μL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16873915
View Documents
Proteintech
25334-1-AP-150UL
150 μL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The HSD17B1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human HSD17B1. This antibody recognizes human antigen. The HSD17B1 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.

Specifications

Specifications

HSD17B1
Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA
Unconjugated
Human
IgG
200 μg/mL
Polyclonal
Rabbit
3292
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.