missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-4 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 300-65-20UG
1813.50 SEK valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.Specifications
Q8WTQ1 | |
protein with no preservative | |
6 | |
20 μg | |
-20°C | |
<1 EU/μg | |
DEFB104A | |
Unconjugated | |
EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
140596 | |
Human BD-4 | |
E. coli | |
RUO | |
BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104 A; Defensin, beta 104; defensin, beta 104 A; defensin, beta 4; hBD-4 | |
Human | |
Recombinant | |
Lyophilized |