missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human BD-4 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
818.00 SEK - 30750.00 SEK
Specifications
Accession Number | Q8WTQ1 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 140596 |
Molecular Weight (g/mol) | 6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Marque | Quantity | Prix | Quantité et disponibilité | |||||
17882733
|
Gibco™
300-65-100UG |
100 μg |
6150.00 SEK
100µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17842803
|
Gibco™
300-65-1MG |
1 mg |
30750.00 SEK
1mg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17872033
|
Gibco™
300-65-20UG |
20 μg |
2015.00 SEK
20µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17852803
|
Gibco™
300-65-250UG |
250 μg |
10770.00 SEK
250µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17871993
|
Gibco™
300-65-500UG |
500 μg |
18430.00 SEK
500µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17842443
|
Gibco™
300-65-5UG |
5 μg |
818.00 SEK
5µg |
Estimated Shipment: 23-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Defensins form a family of antimicrobial and cytotoxic peptides made by neutrophils. Defensins are short, processed peptide molecules that are classified by structure into three groups: alpha-defensins, beta-defensins and theta-defensins. All beta-defensin genes are densely clustered in four to five syntenic chromosomal regions. Chromosome 8p23 contains at least two copies of the duplicated beta-defensin cluster. This duplication results in two identical copies of defensin, beta 104, DEFB104A and DEFB104B, in head-to-head orientation. This gene, DEFB104A, represents the more centromeric copy.Spécification
Q8WTQ1 | |
protein with no preservative | |
6 | |
E. coli | |
RUO | |
BD-4; Beta-defensin 104; Beta-defensin 4; DEFB104; DEFB104A; DEFB104B; DEFB4; DEFB-4; defensin beta 104 A; Defensin, beta 104; defensin, beta 104 A; defensin, beta 4; hBD-4 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
140596 | |
Human BD-4 | |
-20°C | |
<1 EU/μg | |
DEFB104A | |
Unconjugated | |
EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |