Learn More
STEMCELL Technologies Human Recombinant FGF-6, ACF
Fibroblast growth factor 6, animal component-free
Brand: STEMCELL Technologies 78185
This item is not returnable.
View return policy
Description
- Fibroblast growth factor 6 (FGF-6) is a heparin-binding member of the FGF family, which are regulators of cell proliferation, differentiation, and function
- FGF-6 binds and signals through the FGF receptors 1c, 2c, and 4 (Ornitz et al.) FGF-6 is a potent mitogen for fibroblasts, vascular endothelial cells, and prostate carcinoma cells (Asada et al.; Pizette et al.; Ropiquet et al.) FGF-6 is primarily expressed in epithelial and mesenchymal cell lineages
- In development, FGF-6 is expressed in skeletal muscle, consistent with its role in muscle differentiation and regeneration (Floss et al.) FGF-6 has also been shown to promote chondrogenesis in embryonic somites in conjunction with transforming growth factor beta 2 (TGF-β2; Grass et al.) This product is animal component-free
Specifications
Fibroblast growth factor 6, HBGF-6, Heparin-binding growth factor 6, Heparin secretory-transforming protein 2, HST-2, HSTF-2 | |
25 μg | |
Endothelial Cells; Fibroblasts; Mesenchymal Stem and Progenitor Cells; Prostate Cells | |
≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.