missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant FGF-6, ACF
Fibroblast growth factor 6, animal component-free
POA
Specifications
Name | Fibroblast growth factor 6, HBGF-6, Heparin-binding growth factor 6, Heparin secretory-transforming protein 2, HST-2, HSTF-2 |
---|---|
Cell Type | Endothelial Cells; Fibroblasts; Mesenchymal Stem and Progenitor Cells; Prostate Cells |
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109781
|
STEMCELL Technologies
78185.2 |
1000 μg |
POA
1000µg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17189771
|
STEMCELL Technologies
78185 |
25 μg |
POA
25µg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17199771
|
STEMCELL Technologies
78185.1 |
100 μg |
POA
100µg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Fibroblast growth factor 6 (FGF-6) is a heparin-binding member of the FGF family, which are regulators of cell proliferation, differentiation, and function
- FGF-6 binds and signals through the FGF receptors 1c, 2c, and 4 (Ornitz et al.) FGF-6 is a potent mitogen for fibroblasts, vascular endothelial cells, and prostate carcinoma cells (Asada et al.; Pizette et al.; Ropiquet et al.) FGF-6 is primarily expressed in epithelial and mesenchymal cell lineages
- In development, FGF-6 is expressed in skeletal muscle, consistent with its role in muscle differentiation and regeneration (Floss et al.) FGF-6 has also been shown to promote chondrogenesis in embryonic somites in conjunction with transforming growth factor beta 2 (TGF-β2; Grass et al.) This product is animal component-free
Specifications
Fibroblast growth factor 6, HBGF-6, Heparin-binding growth factor 6, Heparin secretory-transforming protein 2, HST-2, HSTF-2 | |
Endothelial Cells; Fibroblasts; Mesenchymal Stem and Progenitor Cells; Prostate Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title