Learn More
Brady™ i5300 Industrial Label Printer Kit EU with Workstation LAB Suite
Set up, switch out and print faster than you ever imagined with the BradyPrinter i5300 Industrial Label Printer. With a solid metal frame, this printer can handle both high-volume and high-mix labeling applications.
Brand:  Brady™ 310852
 View more versions of this product
Product Code. 17449102
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
The i5300 Industrial Label Printer pauses, resumes and stores print jobs in an adjustable queue with overwrite protection.
  • Prints small fonts with precision
  • Requires no calibration and no heat or speed setting adjustments
  • 20 second material changeovers, works with 5.1 to 101.6 mm label widths
  • 178 mm color touchscreen and built-in auto cutter
  • Solid construction with all metal frame
  • Intuitive and easy to operate
  • Includes EU power cord and Brady™ Workstation LAB Suite
Specifications
Specifications
Color LCD Touchscreen
11 in.
11 in.
16 in.
Not Autoclavable
Monochrome
Indonesian, Bulgarian, Chinese (Tradditional), Chinese (Simplified), Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Japanese, Norwegian, Polish, Portuguese, Romanian, Russian, Slovakian, Spanish, Turkish
i5300
600 dpi
Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
33 lb.
Wire and Cable Labeling, Electrical, Laboratory and Barcode Labeling, General Identification
110/240 V
Direct Thermal, Thermal Transfer
284.5 mm
279.4 mm
401 mm
Non-sterile
i5300 Industrial Printer, Brady™ WorkStation Laboratory Identification Suite, EU Power Cable, USB Cable, Stylus, Cleaning Kit, QuickStart Guide, Jump Drive with Videos and 3rd Party Windows™ Driver
512 MB RAM, 195 MB IFFS
254 mm/s
Cabled, Ethernet, USB 2.0, USB Host
10°C to 40°C
15 kg
2 Year
I5300-600-EU-LAB