missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG light chain (lambda) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20758-1-AP-20UL
This item is not returnable.
View return policy
Description
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.Specifications
IgG light chain (lambda) | |
Polyclonal | |
Unconjugated | |
IGLC1 | |
IGLC, IGLC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3537 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0CG04 | |
IGLC1 | |
IgG light chain (lambda) Fusion Protein Ag14693 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |