missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG light chain (lambda) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Specifications
Antigen | IgG light chain (lambda) |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin′s lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore maligt.Specifications
IgG light chain (lambda) | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P0CG04 | |
3537 | |
IgG light chain (lambda) Fusion Protein Ag14693 | |
Primary | |
-20°C | |
IGLC1 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IGLC, IGLC1 | |
IGLC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |