Learn More
IL1RA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10844-1-AP-20UL
This item is not returnable.
View return policy
Description
IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.Specifications
IL1RA | |
Polyclonal | |
Unconjugated | |
IL1RN | |
Anakinra, ICIL 1RA, IL 1ra, IL 1ra3, IL 1RN, IL1 inhibitor, IL1F3, IL1RA, IL1RN, IRAP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3557 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P18510 | |
IL1RN | |
IL1RA Fusion Protein Ag1277 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |