missing translation for 'onlineSavingsMsg'
Learn More

IL1RA Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen IL1RA
Concentration 0.29 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16837503
View Documents Promotion Details
Proteintech
10844-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16827503
View Documents Promotion Details
Proteintech
10844-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL1RA (IL1RN) protein is a cytokine receptor that belongs to the interleukin 1 receptor family. IL1RA is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.
Specifications

Specifications

IL1RA
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human
P18510
3557
IL1RA Fusion Protein Ag1277
Primary
-20°C
IL1RN
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Anakinra, ICIL 1RA, IL 1ra, IL 1ra3, IL 1RN, IL1 inhibitor, IL1F3, IL1RA, IL1RN, IRAP
IL1RN
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications