Learn More
Imidazo[1,2-a]pyridine-8-carboxylic acid hydrochloride, 97%, Thermo Scientific™
1859.06 SEK - 6450.00 SEK
Chemical Identifiers
CAS | 145335-90-6 |
---|---|
Molecular Formula | C8H6N2O2·ClH |
Molecular Weight (g/mol) | 198.61 |
InChI Key | XRQZBGADESKIBB-UHFFFAOYSA-N |
Synonym | imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr |
PubChem CID | 11715356 |
IUPAC Name | imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride |
SMILES | C1=CN2C=CN=C2C(=C1)C(=O)O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||||
11906711
|
Thermo Scientific Acros
440660010 |
1 g | Glass bottle |
1859.06 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11916711
|
Thermo Scientific Acros
440660050 |
5 g | Glass bottle | N/A | N/A | N/A | |||||
Description
Chemical Identifiers
145335-90-6 | |
198.61 | |
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr | |
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride |
C8H6N2O2·ClH | |
XRQZBGADESKIBB-UHFFFAOYSA-N | |
11715356 | |
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl |
Specifications
145335-90-6 | |
White | |
Authentic | |
C8H6N2O2·ClH | |
XRQZBGADESKIBB-UHFFFAOYSA-N | |
imidazo[1,2-a]pyridine-8-carboxylic acid;hydrochloride | |
11715356 | |
97% | |
Imidazo[1, 2-a]pyridine-8-carboxylic acid hydrochloride |
298°C to 300.1°C | |
46.44 to 50.32% C, 13.54 to 14.67% N | |
97% | |
imidazo 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid, monohydrochloride, imidazo 1,2-a pyridine-8-carboxylic acid monohydrochloride, imidazo 1,2-a pyridine-8-carboxylicacidhydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid hydrochloride, imidazo 1,2-alpha pyridine-8-carboxylic acid hydrochloride, imidazol 1,2-a pyridine-8-carboxylic acid monohydr | |
C1=CN2C=CN=C2C(=C1)C(=O)O.Cl | |
198.61 | |
198.61 | |
Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Imidazo¢1,2-a!pyridine-8-carboxylic acid hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.