missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific™
313.65 SEK - 1940.15 SEK
Chemical Identifiers
CAS | 766-55-2 |
---|---|
Molecular Formula | C6H5N3 |
Molecular Weight (g/mol) | 119.12 |
InChI Key | VTVRXITWWZGKHV-UHFFFAOYSA-N |
Synonym | imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene |
PubChem CID | 136599 |
IUPAC Name | imidazo[1,2-b]pyridazine |
SMILES | C1=CC2=NC=CN2N=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15395028
|
Thermo Scientific Acros
455660010 |
1 g |
313.65 SEK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15305038
|
Thermo Scientific Acros
455660050 |
5 g |
564.56 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15315038
|
Thermo Scientific Acros
455660250 |
25 g | N/A | ||||||
Description
Chemical Identifiers
766-55-2 | |
119.12 | |
imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene | |
imidazo[1,2-b]pyridazine |
C6H5N3 | |
VTVRXITWWZGKHV-UHFFFAOYSA-N | |
136599 | |
C1=CC2=NC=CN2N=C1 |
Specifications
766-55-2 | |
Authentic | |
Glass Bottle | |
imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene | |
VTVRXITWWZGKHV-UHFFFAOYSA-N | |
imidazo[1,2-b]pyridazine | |
136599 | |
98% |
54.0°C | |
97.5% min. (GC) | |
C6H5N3 | |
Solubility in water: .. Other solubilities: soluble in dmf | |
C1=CC2=NC=CN2N=C1 | |
119.12 | |
119.12 | |
Imidazo[1,2-b]pyridazine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Imidazo¢1,2-b!pyridazine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only