missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar
Made from borosilicate glass. Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar is perfect for adding precise chemical or liquid that needs to go into a solution. You can combine the required reagents in precise quantities to make a solution.
Brand: Lenz Laborglasinstrumente™ 03026049
This item is not returnable.
View return policy
Description
- With hollow hexagonal stopper NS 29/32
- With scale
- With inscription label
- Material: DURAN™
Specifications
250 mL | |
Conical Ground Glass Joint | |
NS 29/32 | |
85 mm |
Borosilicate Glass | |
Conical | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction