Learn More
Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar
Made from borosilicate glass. Lenz Laborglasinstrumente™ Iodine Determination Flasks with Conical Ground Joint, without Collar is perfect for adding precise chemical or liquid that needs to go into a solution. You can combine the required reagents in precise quantities to make a solution.
181.00 SEK - 2400.00 SEK
Specifications
Material Borosilicate Glass
Includes Conical Ground Glass Joint
Shape Conical
Standard Taper Joint NS 29/32
Graduated Yes
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10080281
View Documents
Lenz Laborglasinstrumente™
03026049
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10599961
View Documents
Lenz Laborglasinstrumente™
03026058
500 mL
2400.00 SEK
Pack of 10
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
Description
Description
  • With hollow hexagonal stopper NS 29/32
  • With scale
  • With inscription label
  • Material: DURAN™
Specifications
Specifications
Borosilicate Glass
Conical
Yes
Conical Ground Glass Joint
NS 29/32