missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

649.00 SEK - 1750.00 SEK

Chemical Identifiers

CAS 18144-43-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017108
InChI Key JWCPZKNBPMSYND-UHFFFAOYSA-N
Synonym isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
PubChem CID 87470
IUPAC Name propan-2-yl 4-aminobenzoate
SMILES CC(C)OC(=O)C1=CC=C(C=C1)N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498177
View Documents
Thermo Scientific Alfa Aesar
25 g
649.00 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
11408187
View Documents
Thermo Scientific Alfa Aesar
100 g
1750.00 SEK
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18144-43-9
179.219
JWCPZKNBPMSYND-UHFFFAOYSA-N
87470
CC(C)OC(=O)C1=CC=C(C=C1)N
C10H13NO2
MFCD00017108
isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
propan-2-yl 4-aminobenzoate

Specifications

18144-43-9
MFCD00017108
JWCPZKNBPMSYND-UHFFFAOYSA-N
propan-2-yl 4-aminobenzoate
87470
98%
C10H13NO2
isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
CC(C)OC(=O)C1=CC=C(C=C1)N
179.219
179.21
Isopropyl 4-aminobenzoate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only