Learn More
Tocris Bioscience™ Ivabradine hydrochloride
HCN channel blocker; inhibits If pacemaker current
Brand: Tocris Bioscience™ 6542/50
2367.11 SEK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Ivabradine hydrochloride is a HCN channel blocker (IC50 is approximately 0.5 - 2.5 μM). Bradycardic agent that inhibits If pacemaker current in sinoatrial node cells.Specifications
3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride | |
50 mg | |
98% |
148849-67-6 | |
C27H36N2O5.HCl |