missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ivabradine hydrochloride
HCN channel blocker; inhibits If pacemaker current
787.24 SEK - 2958.89 SEK
Specifications
Chemical Name or Material | 3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride |
---|---|
CAS | 148849-67-6 |
Molecular Formula | C27H36N2O5.HCl |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18767596
|
Tocris Bioscience™
6542/10 |
10 mg |
787.24 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18706326
|
Tocris Bioscience™
6542/50 |
50 mg |
2958.89 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HCN channel blocker (IC50 is approximately 0.5 - 2.5 μM). Bradycardic agent that inhibits If pacemaker current in sinoatrial node cells.Specifications
3-[3-[[[(7S)-3,4-Dimethoxybicyclo[4.2.0]octa-1,3,5-trien-7-yl]methyl]methylamino]propyl]-1,3,4,5-tetrahydro-7,8-dimethoxy-2H-3-benzazepin-2-one hydrochloride | |
148849-67-6 | |
C27H36N2O5.HCl | |
98% |
Safety and Handling
Recommended Storage : Store at -20°C