Learn More
Justrite™ Safety Fill Vent
Brand:  Justrite™ 08204
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15173607
  • 1610.00 SEK / Each
Estimated Shipment: 09-09-2024
to see stock.
Specifications
Fill Drum Vent
Facilitate Waste Solvent Disposal
6 in. Flame arrester, automatic pressure and vacuum relief and cap with chain
FM
Brass