man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Facility Safety and Maintenance
Hazardous Materials Storage
Drums and Accessories
Drum Accessories
Justrite™ Safety Fill Vent
Print
Justrite™ Safety Fill Vent
1610.00 SEK
See promotional offers below
Specifications
Product Type
Fill Drum Vent
Certifications/Compliance
FM
View More Specs
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
Includes
Price
Quantity & Availability
Product Code
Brand
Includes
Price
Quantity & Availability
15173607
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Justrite™
08204
6 in. Flame arrester, automatic pressure and vacuum relief and cap with chain
1610.00 SEK
Each
View alternative products
Save Up to
»
Estimated Shipment: 10-09-2024
Log in
to see stock.
Specifications
Product Type
Fill Drum Vent
Certifications/Compliance
FM
Documents
Product Certifications
Back to Top