missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMO Mouse anti-Human, Mouse, Rat, Clone: 2D2F2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60029-1-IG-150UL
This item is not returnable.
View return policy
Description
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.Specifications
KMO | |
Monoclonal | |
0.62 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15229, O88867, Q91WN4 | |
Kmo | |
KMO Fusion Protein Ag1109 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2D2F2 | |
Unconjugated | |
KMO | |
KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
59113, 8564, 98256 | |
-20°C | |
Liquid |