missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KMO Mouse anti-Human, Mouse, Rat, Clone: 2D2F2, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KMO |
---|---|
Clone | 2D2F2 |
Concentration | 0.62 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816985
|
Proteintech
60029-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806985
|
Proteintech
60029-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kynurenine 3-monooxygenase (KMO; EC 1.14.13.9) is an NADPH-dependent flavin monooxygenase that catalyzes the hydroxylation of the L-tryptophan metabolite L-kynurenine to form L-3-hydroxykynurenine.Specifications
KMO | |
0.62 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KMO, Kynurenine 3 hydroxylase, Kynurenine 3 monooxygenase | |
Kmo | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
2D2F2 | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat, Mouse | |
O15229, O88867, Q91WN4 | |
59113, 8564, 98256 | |
KMO Fusion Protein Ag1109 | |
Primary | |
-20°C | |
KMO |