missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ornithine Hydrochloride, 99%, MP Biomedicals
L-Ornithine is a product of arginase degradation and plays an important role in ammonia metabolism via the urea cycle
605.00 SEK - 46320.00 SEK
Chemical Identifiers
CAS | 3184-13-2 |
---|---|
Molecular Formula | C5H13ClN2O2 |
Molecular Weight (g/mol) | 168.621 |
MDL Number | MFCD00064562 |
InChI Key | GGTYBZJRPHEQDG-WCCKRBBISA-N |
Synonym | l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate |
PubChem CID | 76654 |
IUPAC Name | (2S)-2,5-diaminopentanoic acid;hydrochloride |
SMILES | C(CC(C(=O)O)N)CN.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11471961
|
MP Biomedicals
0219471825 |
25 g |
605.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11481961
|
MP Biomedicals
0219471880 |
100 g |
2065.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413100
|
MP Biomedicals
0210042194 |
5 kg |
46320.00 SEK
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
3184-13-2 | |
168.621 | |
GGTYBZJRPHEQDG-WCCKRBBISA-N | |
76654 | |
C(CC(C(=O)O)N)CN.Cl |
C5H13ClN2O2 | |
MFCD00064562 | |
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
(2S)-2,5-diaminopentanoic acid;hydrochloride |
Specifications
3184-13-2 | |
MFCD00064562 | |
GGTYBZJRPHEQDG-WCCKRBBISA-N | |
(2S)-2,5-diaminopentanoic acid;hydrochloride | |
76654 | |
Cell Culture Reagent | |
L-Ornithine, 99%, Hydrochloride, This material is essentially free of citrulline and ammonia. |
C5H13ClN2O2 | |
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
C(CC(C(=O)O)N)CN.Cl | |
168.621 | |
168.6 | |
Crystalline |
Safety and Handling
Recommended Storage : Room temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title