Learn More
Hirschmann™ labopette™ fix
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand:  Hirschmann™ 9475513
 View more versions of this product
Product Code. 11632269
  • 2925.00 SEK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Fixed volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Specifications
0.6%
Clear: 1-10 mL
Autoclavable
Manual Pipette
Calibration device, individual number and weighing protocol
1 Each
ISO 8655
Tip ejector
1
10 mL
0.3%