Learn More
Hirschmann™ labopette™ fix
Ergonomically shaped manual pipette, effortless operation and tip ejector.
2925.00 SEK
Specifications
Certifications/Compliance | ISO 8655 |
---|---|
Description | Tip ejector |
Autoclavable | Autoclavable |
Number of Channels | 1 |
Type | Manual Pipette |
Product Code | Brand | Volume (Metric) | Accuracy | Compatible Tips | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Accuracy | Compatible Tips | Price | Quantity & Availability | |||
11612259
|
Hirschmann™
9475501 |
5 μL | 2.5% | Clear: 0.2-10 μL, yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11622259
|
Hirschmann™
9475502 |
10 μL | 1.2% | Clear: 0.2-10 μL, yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11632259
|
Hirschmann™
9475503 |
20 μL | 1.0% | Yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11642259
|
Hirschmann™
9475504 |
25 μL | 1.0% | Yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11652259
|
Hirschmann™
9475505 |
50 μL | 1.0% | Yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11662259
|
Hirschmann™
9475506 |
100 μL | 0.8% | Yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11672259
|
Hirschmann™
9475507 |
200 μL | 0.8% | Yellow: 1-200 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11682259
|
Hirschmann™
9475508 |
250 μL | 0.8% | Blue: 10-1000 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11692259
|
Hirschmann™
9475509 |
500 μL | 0.8% | Blue: 10-1000 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11602269
|
Hirschmann™
9475510 |
1000 μL | 0.8% | Blue: 10-1000 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11612269
|
Hirschmann™
9475511 |
2000 μL | 0.8% | Clear: 100-5000 μL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11632269
|
Hirschmann™
9475513 |
10 mL | 0.6% | Clear: 1-10 mL |
2925.00 SEK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Fixed volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
ISO 8655 | |
Autoclavable | |
Manual Pipette | |
1 Each |
Tip ejector | |
1 | |
Calibration device, individual number and weighing protocol |