Learn More
Hirschmann™ labopette™, Variable
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand: Hirschmann™ 9475400
This item is not returnable.
View return policy
Description
- Variable volume setting
- With tip ejector and recalibration possibility
- Autoclavable
Specifications
4.0% | |
Clear, 0.2-10 μL | |
Autoclavable | |
Labopette™ | |
0.1 to 3 μL | |
0.002 μL | |
1 Each |
ISO 8655 | |
Tip ejector | |
1 | |
Variable Volume Pipette | |
Calibration device, individual number and weighing protocol | |
2.0% |