Learn More
Hirschmann™ labopette™, Variable
Ergonomically shaped manual pipette, effortless operation and tip ejector.
4845.00 SEK
Specifications
Certifications/Compliance ISO 8655
Description Tip ejector
Autoclavable Autoclavable
Number of Channels 1
Product Line Labopette™
View More Specs
Products 4
Product Code Brand Volume (Metric) Accuracy Compatible Tips Increments Price Quantity & Availability  
Product Code Brand Volume (Metric) Accuracy Compatible Tips Increments Price Quantity & Availability  
11672249
View Documents
Hirschmann™
9475400
0.1 to 3 μL 4.0% Clear, 0.2-10 μL 0.002 μL
4845.00 SEK
Each
10109664
View Documents
Hirschmann™
9475401
0.5 to 10 μL 1.2% Clear: 0.2-10 μL, yellow: 1-200 μL 0.01 μL
4845.00 SEK
Each
11662249
View Documents
Hirschmann™
9475404
1 to 10 mL 0.6% Clear: 1-10 mL 20 μL
4845.00 SEK
Each
11682249
View Documents
Hirschmann™
9475406
2 to 20 μL 1.0% Yellow, 1-200 μL 0.02 μL
4845.00 SEK
Each
Description
Description
  • Variable volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Specifications
ISO 8655
Autoclavable
Labopette™
Calibration device, individual number and weighing protocol
Tip ejector
1
Variable Volume Pipette
1 Each