Learn More
Hirschmann™ labopette™, Variable
Ergonomically shaped manual pipette, effortless operation and tip ejector.
Brand:  Hirschmann™ 9475406
 View more versions of this product
Product Code. 11682249
  • 4845.00 SEK / Each
Explore more special offers
Description
Description
  • Variable volume setting
  • With tip ejector and recalibration possibility
  • Autoclavable
Specifications
Specifications
1.0%
Yellow, 1-200 μL
Autoclavable
Labopette™
2 to 20 μL
0.02 μL
1 Each
ISO 8655
Tip ejector
1
Variable Volume Pipette
Calibration device, individual number and weighing protocol
0.5%